RABBIT IgG FRACTION TO HUMAN IgG (WHOLE MOLECULE). Anti-HumanHost: rabbitForm: lyophilized (IgG)Applications: Immunostaining of acet one-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 2 mL
Catalog # ICNA0855008
Supplier: MP Biomedicals
Frequently Bought Together