Order Entry
Netherlands
ContactUsLinkComponent
FE:PB:CA:CU: o.d. (260nm) of a saturated solution. Useful in the isolation and purification of enzymes and proteins. Supplied as a white, crystalline solid. 1 * 50 g
Catalog # ICNA04808210
Supplier:  MP Biomedicals
FE:PB:CA:CU: o.d. (260nm) of a saturated solution. Useful in the isolation and purification of enzymes and proteins. Supplied as a white, crystalline solid. 1 * 50 g
Catalog # ICNA04808210
Supplier:  MP Biomedicals
Frequently Bought Together