Order Entry
Netherlands
ContactUsLinkComponent
FITC CONJUGATED GOAT AFFINITY PURIFIED IgG TO DOG IgG (WHOLE MOLECULE). Anti-Dog Host: goat Form: affinity purified liquid Applicati ons: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. FITC Conjugated 1 * 2 mg
Catalog # ICNA0856998
Supplier:  MP Biomedicals
FITC CONJUGATED GOAT AFFINITY PURIFIED IgG TO DOG IgG (WHOLE MOLECULE). Anti-Dog Host: goat Form: affinity purified liquid Applicati ons: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. FITC Conjugated 1 * 2 mg
Catalog # ICNA0856998
Supplier:  MP Biomedicals
Frequently Bought Together