2-Phenoxyethanol
100 mL
Catalog # ICNA0215014880
Supplier: MP Biomedicals
Jump to:
Product Details & Documents
Important notice: this product belongs to MP Biomedicals rare chemical library. Most rare chemical products are available for immediate delivery and sold "As Is". MP biomedicals does not collect analytical data for those products. All sales are final. However, analytical test may be performed upon custom request for bulk orders.
: Research Use Only (RUO).
Synonyms: ω-Hydroxyphenetole, β-Hydroxyphenetole, Phenyl cellosolve, Phenoxyethanol, Ethylene glycol monophenyl ether
- CAS Number:122-99-6
- Formula:C₆H₅OCH₂CH₂OH
- MDL Number:MFCD00002857
- Molecular Weight:138.17 g/mol
- Melting Pt:57 ° F (lit.)
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
Specifications
- Pk:100 mL



