Order Entry
Netherlands
ContactUsLinkComponent
Crystalline Purity: 97% Used as a non-ionic detergent for stabilization and activation of enzymes and for membrane research. 1 * 5 g
Catalog # ICNA0215101394
Supplier:  MP Biomedicals
Crystalline Purity: 97% Used as a non-ionic detergent for stabilization and activation of enzymes and for membrane research. 1 * 5 g
Catalog # ICNA0215101394
Supplier:  MP Biomedicals
Frequently Bought Together