Order Entry
Netherlands
ContactUsLinkComponent
APROTININ IN SOLUTION ANALYTICAL GRADE. From Bovine LungAnalytical GradeAseptically filled solution in 0.9% NaCl and 0.9% benzyl alc ohol.Activity: 100,000 KIU/10 ml ampule 1 * 40 mL
Catalog # ICNA0219077940
Supplier:  MP Biomedicals
APROTININ IN SOLUTION ANALYTICAL GRADE. From Bovine LungAnalytical GradeAseptically filled solution in 0.9% NaCl and 0.9% benzyl alc ohol.Activity: 100,000 KIU/10 ml ampule 1 * 40 mL
Catalog # ICNA0219077940
Supplier:  MP Biomedicals
Frequently Bought Together