bis - TRIS. Cell Culture Reagent.Purity: 99+% Zwitterionic buffer pKa = 6.5 at 25C Useful pH range 5.8-7.2. 1 * 25 g
Catalog # ICNA0219454625
Supplier: MP Biomedicals
bis - TRIS. Cell Culture Reagent.Purity: 99+% Zwitterionic buffer pKa = 6.5 at 25C Useful pH range 5.8-7.2. 1 * 25 g
Catalog # ICNA0219454625
Supplier: MP Biomedicals
Frequently Bought Together
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection