Order Entry
Netherlands
ContactUsLinkComponent
Off-white to yellow powder.Reagent for fluorescent assay of amino acids, peptides, proteins and other primary amines. 1,2. Also used for fluorometric assays of some proteolytic enzymes.3 1 * 250 mg
Catalog # ICNA0219518383
Supplier:  MP Biomedicals
Off-white to yellow powder.Reagent for fluorescent assay of amino acids, peptides, proteins and other primary amines. 1,2. Also used for fluorometric assays of some proteolytic enzymes.3 1 * 250 mg
Catalog # ICNA0219518383
Supplier:  MP Biomedicals
Frequently Bought Together