MELANGE ACETONE/EAU AVEC BLEU DE BROMOPHENOL 0,02G/L 1 * 5 L
Catalog # PRLS89617.360
Supplier: PROLABO CMP
MELANGE ACETONE/EAU AVEC BLEU DE BROMOPHENOL 0,02G/L 1 * 5 L
Catalog # PRLS89617.360
Supplier: PROLABO CMP
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Kieselguhr, AnalaR NORMAPUR®, purified and calcined
Supplier: VWR Chemicals
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Test kits, nitrite, Spectroquant®, Supelco®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
Supplier: VWR Collection
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
VWR® Rota®, pH indicator paper, 1 to 14
Supplier: VWR Chemicals