SPIROCHETE TREPONEMA PALLIDUM 1 * 6 mL
Catalog # BCARPP135AA
Supplier: BIOCARE MEDICAL
Frequently Bought Together
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Acetic acid glacial ≥98%, TECHNICAL
Supplier: VWR Chemicals
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Ammonia solution 25%, LiChropur™ for HPLC, Supelco®
Supplier: Merck
Test strips for arsenic, MQuant®, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Test kits, chloride, Spectroquant®, Supelco®
Supplier: Merck
Starch indicator solution, MDB
Supplier: Hach
Test kits, photometric, ammonium, LCK cuvette tests
Supplier: Hach