Order Entry
Netherlands
ContactUsLinkComponent
An inhibitor of PARPs which can be used in cells or in animals 1 * 10 mg
Catalog # CAYM14440-10
Supplier:  Cayman Chemical
An inhibitor of PARPs which can be used in cells or in animals 1 * 10 mg
Catalog # CAYM14440-10
Supplier:  Cayman Chemical
Frequently Bought Together