Order Entry
Netherlands
ContactUsLinkComponent
A quinolone antibiotic that inhibits bacterial DNA gyrase, but not eukaryotic topoisomerases, reversibly binding gyrase subunit A in gyrase-DNA complexes , blocking supercoiling activity and inhibiting DNA synthesis at 0.5-5 µg/ml 1 * 10 g
Catalog # CAYM16789-10
Supplier:  Cayman Chemical
A quinolone antibiotic that inhibits bacterial DNA gyrase, but not eukaryotic topoisomerases, reversibly binding gyrase subunit A in gyrase-DNA complexes , blocking supercoiling activity and inhibiting DNA synthesis at 0.5-5 µg/ml 1 * 10 g
Catalog # CAYM16789-10
Supplier:  Cayman Chemical
Frequently Bought Together