Order Entry
Netherlands
ContactUsLinkComponent
An antibiotic that can inhibit cell wall synthesis in plant-pathogenic oomycetes (MIC = 25 µg/ml for P. medicaginis M2913) and certa in fungi as well as some bacterial species (MIC = 400 µg/ml for S. aureus) 1 * 5 mg
Catalog # CAYM16623-5
Supplier:  Cayman Chemical
An antibiotic that can inhibit cell wall synthesis in plant-pathogenic oomycetes (MIC = 25 µg/ml for P. medicaginis M2913) and certa in fungi as well as some bacterial species (MIC = 400 µg/ml for S. aureus) 1 * 5 mg
Catalog # CAYM16623-5
Supplier:  Cayman Chemical
Frequently Bought Together