Order Entry
Netherlands
ContactUsLinkComponent
A selective inhibitor of the 5i (LMP7) subunit of the immunoproteasome (IC50s = 65 and 73 nM for mouse and human, respectively) tha t demonstrates significantly weaker activity at the 5 subunit of the constitutive proteasome (IC50s = 0.92 and 1.04 1 * 10 mg
Catalog # CAYM16271-10
Supplier:  Cayman Chemical
A selective inhibitor of the 5i (LMP7) subunit of the immunoproteasome (IC50s = 65 and 73 nM for mouse and human, respectively) tha t demonstrates significantly weaker activity at the 5 subunit of the constitutive proteasome (IC50s = 0.92 and 1.04 1 * 10 mg
Catalog # CAYM16271-10
Supplier:  Cayman Chemical
Frequently Bought Together