A potent, selective agonist of the neuromedin K3 (NK3) receptor (EC50 = 0.5-3 nM) 1 * 500 µG
CAYM16721-500
A potent, selective agonist of the neuromedin K3 (NK3) receptor (EC50 = 0.5-3 nM) 1 * 500 µG
CAYM16721-500
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection