Order Entry
Netherlands
ContactUsLinkComponent
An analog of dapagliflozin that displays similar selectivity for inhibiting SGLT2 over SGLT1 (Kis = 5.3 and 330 nM, respectively) 1 * 10 mg
Catalog # CAYM15951-10
Supplier:  Cayman Chemical
An analog of dapagliflozin that displays similar selectivity for inhibiting SGLT2 over SGLT1 (Kis = 5.3 and 330 nM, respectively) 1 * 10 mg
Catalog # CAYM15951-10
Supplier:  Cayman Chemical
Frequently Bought Together