Inhibits the p53-Mdm2 interaction (Ki = 83 nM) and also dose-dependently reduces activation of the NF-B pathway 1 * 5 mg
CAYM15574-5
Inhibits the p53-Mdm2 interaction (Ki = 83 nM) and also dose-dependently reduces activation of the NF-B pathway 1 * 5 mg
CAYM15574-5
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection