Order Entry
Netherlands
ContactUsLinkComponent
A fluorescent probe that can react with ONOO-, HOCl, and H2O2 in a similar manner to CBA, yet is formulated for improved solubility 1 * 5 mg
Catalog # CAYM10818-5
Supplier:  Cayman Chemical
A fluorescent probe that can react with ONOO-, HOCl, and H2O2 in a similar manner to CBA, yet is formulated for improved solubility 1 * 5 mg
Catalog # CAYM10818-5
Supplier:  Cayman Chemical
Frequently Bought Together