Order Entry
Netherlands
ContactUsLinkComponent
A CPT1 and CPT2 inhibitor that inhibits rat heart and liver CPT1 (IC50 = 77 and 148 µM, respectively), and rat heart CPT2 (IC50 = 79 µM) 1 * 10 mg
Catalog # CAYM16982-10
Supplier:  Cayman Chemical
A CPT1 and CPT2 inhibitor that inhibits rat heart and liver CPT1 (IC50 = 77 and 148 µM, respectively), and rat heart CPT2 (IC50 = 79 µM) 1 * 10 mg
Catalog # CAYM16982-10
Supplier:  Cayman Chemical
Frequently Bought Together