Order Entry
Netherlands
ContactUsLinkComponent
A cytidine analog with an unnatural -L structural configuration that inhibits reverse transcriptase, preventing replication of HIV- 1 (EC50 = 10-20 nM) and hepatitis B (EC50 = 10-40 nM) 1 * 250 mg
Catalog # CAYM16879-250
Supplier:  Cayman Chemical
A cytidine analog with an unnatural -L structural configuration that inhibits reverse transcriptase, preventing replication of HIV- 1 (EC50 = 10-20 nM) and hepatitis B (EC50 = 10-40 nM) 1 * 250 mg
Catalog # CAYM16879-250
Supplier:  Cayman Chemical
Frequently Bought Together