Order Entry
Netherlands
ContactUsLinkComponent
An orally active, selective inhibitor of Cdk4 (IC50 = 11 nM) and Cdk6 (IC50 = 16 nM) with antiproliferative activity against retinob lastoma-positive tumor cells, blocking retinoblastoma phosphorylation and inducing G1 arrest 1 * 5 mg
Catalog # CAYM16273-5
Supplier:  Cayman Chemical
An orally active, selective inhibitor of Cdk4 (IC50 = 11 nM) and Cdk6 (IC50 = 16 nM) with antiproliferative activity against retinob lastoma-positive tumor cells, blocking retinoblastoma phosphorylation and inducing G1 arrest 1 * 5 mg
Catalog # CAYM16273-5
Supplier:  Cayman Chemical
Frequently Bought Together