Order Entry
Netherlands
ContactUsLinkComponent
A serotonin 5-HT1B and 5-HT1D receptor agonist (IC50s = 9.3 and 7.3 nM, respectively) that also has affinity for the 5-HT1F receptor (IC50 = 17.8 nM) 1 * 25 mg
Catalog # CAYM14600-25
Supplier:  Cayman Chemical
A serotonin 5-HT1B and 5-HT1D receptor agonist (IC50s = 9.3 and 7.3 nM, respectively) that also has affinity for the 5-HT1F receptor (IC50 = 17.8 nM) 1 * 25 mg
Catalog # CAYM14600-25
Supplier:  Cayman Chemical
Frequently Bought Together