Order Entry
Netherlands
ContactUsLinkComponent
A first generation tricyclic antidepressant that acts primarily as an inhibitor of serotonin and norepinephrine transporters (Kds = 1.4 and 37 nM, respectively) 1 * 5 g
Catalogus # CAYM15890-5
Supplier:  Cayman Chemical
A first generation tricyclic antidepressant that acts primarily as an inhibitor of serotonin and norepinephrine transporters (Kds = 1.4 and 37 nM, respectively) 1 * 5 g
Catalogus # CAYM15890-5
Supplier:  Cayman Chemical
Frequently Bought Together