Order Entry
Netherlands
ContactUsLinkComponent
Rotavapor R-300 System B-305, SJ24/40, C, P+G, I-300P, V-300, F-305, 230V. Inc. V-300 pump, I-300 int. w/o chiller. Elec. lift, B-305 bath, SJ 24/40 Glass set: Cold trap, P+G coating, int.: I-300 Pro, Woulff bottle F-305 chiller 1 * 1 items
Catalog # BUCH11SR300252CP11
Supplier:  BUCHI
Rotavapor R-300 System B-305, SJ24/40, C, P+G, I-300P, V-300, F-305, 230V. Inc. V-300 pump, I-300 int. w/o chiller. Elec. lift, B-305 bath, SJ 24/40 Glass set: Cold trap, P+G coating, int.: I-300 Pro, Woulff bottle F-305 chiller 1 * 1 items
Catalog # BUCH11SR300252CP11
Supplier:  BUCHI
Frequently Bought Together