PHENOLPHTALEINE 1% DANS METHANOL 1 * 250 mL
Catalog # PRLS89572.230
Supplier: PROLABO CMP
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Cryocompound reagents, Immunologic
Supplier: IMMUNOLOGIC
Avantor® Seradigm, Fetal Bovine Serum (FBS), US origin
Supplier: VWR Seradigm
pH indicator papers, universal/special
Supplier: MACHEREY-NAGEL
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
Supplier: Hach
Eosin Y (yellowish), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Reagents for Sulphide Determination using Lovibond® MD photometers
Supplier: Lovibond
Reagents for ammonia determination using Lovibond® MD photometers
Supplier: Lovibond