Order Entry
Netherlands
ContactUsLinkComponent
Rotavapor R-300 B-301, SJ29/32, A, I-300, VU, 230V. . Lift System: Manual Heating Bath: B-301 (20 - 95?, 1L) SJ 29/32 Glass set: Di agonal (A) int.: I-300, valve unit 1 * 1 items
Catalog # BUCH11R300111A041
Supplier:  BUCHI
Rotavapor R-300 B-301, SJ29/32, A, I-300, VU, 230V. . Lift System: Manual Heating Bath: B-301 (20 - 95?, 1L) SJ 29/32 Glass set: Di agonal (A) int.: I-300, valve unit 1 * 1 items
Catalog # BUCH11R300111A041
Supplier:  BUCHI
Frequently Bought Together