Order Entry
Netherlands
ContactUsLinkComponent
Rotavapor R-300 no bath, SJ29/32, V, I-300P, 230V. . Lift System: Manual Heating Bath: w/o SJ 29/32 Glass set: Vertical (V) int.: I -300 Pro, Woulff bottle 1 * 1 items
Catalog # BUCH11R300101V031
Supplier:  BUCHI
Rotavapor R-300 no bath, SJ29/32, V, I-300P, 230V. . Lift System: Manual Heating Bath: w/o SJ 29/32 Glass set: Vertical (V) int.: I -300 Pro, Woulff bottle 1 * 1 items
Catalog # BUCH11R300101V031
Supplier:  BUCHI
Frequently Bought Together