Rotavapor R-300 B-305, SJ24/40, no glass, I-300P, VU, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) SJ 24/40 Glas s set: w/o int.: I-300 Pro, valve unit 1 * 1 items
Catalog # BUCH11R300152N051
Supplier: BUCHI
Frequently Bought Together