Order Entry
Netherlands
ContactUsLinkComponent
Potential-free contact (24 V/2 A) with socket to NAMUR NE 28, for signal generation, controlled by programme segment, for freely sel ectable functions to be activated (e.g. activation of audible and visual signals, exhaust Only for units with TwinDIS 1 * 1 items
Catalog # MEMMUF450TSH72
Supplier:  MEMMERT
Potential-free contact (24 V/2 A) with socket to NAMUR NE 28, for signal generation, controlled by programme segment, for freely sel ectable functions to be activated (e.g. activation of audible and visual signals, exhaust Only for units with TwinDIS 1 * 1 items
Catalog # MEMMUF450TSH72
Supplier:  MEMMERT
Frequently Bought Together