Order Entry
Netherlands
ContactUsLinkComponent
4-20 MA CURRENT LOOP INTERFACE (0 TO +310 °C = 4 - 20 MA) - TEMPERATUR OF A PT100 SENSOR POSITIONED FLEXIBLY IN CHAMBER FOR EXTERNAL TEMPERATURE MONITORING (MAX. 1 SINGLEDISPLAY MAX. 3 TWINDISPLAY) - PRICE PER SENSOR 1 * 1 items
Catalog # MEMMUF160TSV6
Supplier:  MEMMERT
4-20 MA CURRENT LOOP INTERFACE (0 TO +310 °C = 4 - 20 MA) - TEMPERATUR OF A PT100 SENSOR POSITIONED FLEXIBLY IN CHAMBER FOR EXTERNAL TEMPERATURE MONITORING (MAX. 1 SINGLEDISPLAY MAX. 3 TWINDISPLAY) - PRICE PER SENSOR 1 * 1 items
Catalog # MEMMUF160TSV6
Supplier:  MEMMERT
Frequently Bought Together