Morpholine 99+% ACS
Glass bottle, 2,5 kg
- :110-91-8
- :C₄H₉NO
- :MFCD00005972
- :87.12 g/mol
- :12,06362
- :128.9 °C (1013 hPa)
- :–5 °C
- :0.99 g/cm³
- :32 °C
- :Ambient
- :203-815-1
- :2054
- :8,I
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4530 to 1.4550 (20°C, 589 nm)
- Boiling point:126.0 to 130.0°C
- Color scale:≤15 APHA
- Titration with HCl:≥99.0 %
- Pack type:Glass bottle
- Pk:2,5 kg
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



