Borane trimethylamine complex 97%
Glass bottle, 25 g
Supplier: Thermo Fisher Scientific
Borane trimethylamine complex 97%
Catalog # ACRO402740250
Supplier: Thermo Fisher Scientific
CAS Number: 75-22-9
Jump to:
Product Details & Documents
- CAS Number:75-22-9
- Formula:C₃H₁₂BN
- MDL Number:MFCD00012420
- Molecular Weight:72.95 g/mol
- Melting Pt:92…96 °C
- Storage Temperature:Ambient
- EINECS:200-850-4
- UN:1325
- ADR:4.1,II
Documents
ACRO40274
SDS
Specifications
Specification Test Results
- Appearance:White Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:92 to 96 °C
- Titration with Na2S2O3:≥96.0 %
Specifications
- Pack type:Glass bottle
- Pk:25 g



