Propionic anhydride 99%
Glass bottle, 250 mL
Supplier: Thermo Fisher Scientific
Propionic anhydride 99%
Catalog # ACRO131522500
Supplier: Thermo Fisher Scientific
CAS Number: 123-62-6
Jump to:
Product Details & Documents
- CAS Number:123-62-6
- Formula:C₆H₁₀O₃
- MDL Number:MFCD00009303
- Molecular Weight:130.14 g/mol
- Merck Index:12,08011
- Boiling Pt:167 °C (1013 hPa)
- Melting Pt:–42 °C
- Density:1.015 g/cm³
- Flash Pt:63 °C
- Storage Temperature:Ambient
- EINECS:204-638-2
- UN:2496
- ADR:8,III
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4030 to 1.4050 (20°C, 589 nm)
- GC:≥98.5 %
- Color scale:≤20 APHA
Specifications
- Pack type:Glass bottle
- Pk:250 mL



