2,5-Dibromothiophene 95%
Glass bottle, 500 g
- :3141-27-3
- :C₄H₂Br₂S
- :MFCD00005420
- :241.93 g/mol
- :211 °C (760 mmHg)
- :–6 °C
- :2.14 g/cm³
- :99 °C
- :Ambient
- :221-547-3
- :6.1,II
Documents
ACRO14868
SDS
- Appearance:Clear light yellow to brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6270 to 1.6300 (20°C, 589 nm)
- GC:≥94 %
- Pack type:Glass bottle
- Pk:500 g



