Melatonin 99%
Glass bottle, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide
- CAS Number:73-31-4
- Formula:C₁₃H₁₆N₂O₂
- MDL Number:MFCD00005655
- Molecular Weight:232.28 g/mol
- Boiling Pt:512.8 °C
- Melting Pt:118 °C
- Flash Pt:264 °C
- Storage Temperature:Ambient
- EINECS:200-797-7
Documents
Specifications
Specification Test Results
- Appearance:White to cream to yellow Powder
- Infrared spectrum:Conforms
- Melting point:116 to 120 °C
- Loss on drying:≤0.5 % (1 g, 105 °C)
- Heavy metals:≤20 ppm
- HPLC:≥98.5 %
Specifications
- Pack type:Glass bottle
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



