Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Sodium cholate 99%

Sodium cholate 99%

Glass bottle, 500 g
Catalog # ACRO405195000
Irritant
Sodium cholate 99%
Catalog # ACRO405195000
CAS Number:  361-09-1
Sodium cholate 99%
Sodium cholate 99%

Glass bottle, 500 g

CAS Number:  361-09-1
Price
375,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  3α,7α,12α-Trihydroxy-5β-cholanic acid sodium salt, 3α,7α,12α-Trihydroxycholan-24-oic acid sodium salt, 3α,7α,12α-Trihydroxycholanic acid sodium salt, Sodium 3α,7α,12α-trihydroxy-5β-cholanoate, (R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid sodium salt, 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid sodium salt
  • CAS Number:361-09-1
  • Formula:C₂₄H₃₉NaO₅
  • MDL Number:MFCD00150749
  • Molecular Weight:430.56 g/mol
  • Melting Pt:198 °C
  • Storage Temperature:Ambient
  • EINECS:206-643-5

Documents

Specifications

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤5.0 % (110°C)
  • pH:
    7.5 to 9.5 (5 % in water)
  • Specific optical rotation:
    +33° to +37° (20°C, 589 nm) (c=0.6, ethanol)
  • Titration with HClO4:
    98.0 to 102.0 % (On dried basis)
  • Heavy metals (as Pb):
    ≤50 ppm
  • Impurity:
    ≤2.0 % Sodium deoxycholate

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.