Bromelain (juice) (from Stem)
Glass bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Documents
ACRO44821
SDS
Specifications
Specifications
- Pack type:Glass bottle
- Pk:100 g
- Cat. No.:ACRO448211000
- Form:Cream to light brown powder
- Storage conditions:Refrigerator
- MDL No.:MFCD00081423
- Source:Stem
- Enzyme name:Bromelain (juice)
- CAS No.:9001-00-7
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



