CONOSTAN CUSTOM BLEND:S-21+K, 100PPM 1 * 200 g
Catalog # PROOCON150-021-052
Supplier: LGC Standards PROMOCHEM
CONOSTAN CUSTOM BLEND:S-21+K, 100PPM 1 * 200 g
Catalog # PROOCON150-021-052
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals