Order Entry
Netherlands
ContactUsLinkComponent
MicroKros 41,5cm MPES 750kD 1,0mm MLL X FLL dry 1 * 1 items
  SPECC04-E750-10-N
MicroKros 41,5cm MPES 750kD 1,0mm MLL X FLL dry 1 * 1 items
  SPECC04-E750-10-N
Frequently Bought Together