Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Additional Top and Bottom Shelves
Additional Top and Bottom Shelves
Bottom shelf, solid, for cleanroom heavy duty tables, L×W: 1500×300, fabricated from stainless steel 304 with electropolished finish
Catalog # PALBWX1519
Supplier:   Palbam
Additional Top and Bottom Shelves
Catalog # PALBWX1519
Supplier:  Palbam
Additional Top and Bottom Shelves
Additional Top and Bottom Shelves
Bottom shelf, solid, for cleanroom heavy duty tables, L×W: 1500×300, fabricated from stainless steel 304 with electropolished finish
Price
718,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Bottom shelf, solid, for cleanroom heavy duty tables, L×W: 1500×300, fabricated from stainless steel 304 with electropolished finish
  • Seq:
    09