Order Entry
Netherlands
ContactUsLinkComponent
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers 1 * 1 items
Catalog # ALLA50203T150-QP
Supplier:  Alla France
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers 1 * 1 items
Catalog # ALLA50203T150-QP
Supplier:  Alla France
Frequently Bought Together