Order Entry
Netherlands
ContactUsLinkComponent
Concentric nozzle set. Set of 7 external nozzles with high precision opening of 0.2, 0.3, 0.4, 0.5, 0.6, 0.7 and 0.9mm, made of stainless steel, concentric bead producing unit and 1000ml pressure bottle. 1 * 1 SET
Catalog # BUCH11058051
Supplier:  BUCHI
Concentric nozzle set. Set of 7 external nozzles with high precision opening of 0.2, 0.3, 0.4, 0.5, 0.6, 0.7 and 0.9mm, made of stainless steel, concentric bead producing unit and 1000ml pressure bottle. 1 * 1 SET
Catalog # BUCH11058051
Supplier:  BUCHI
Frequently Bought Together