Order Entry
Netherlands
ContactUsLinkComponent
47mm Petri dish with pad, sterile, to be filled with liquid media for microbial culture 1 * 150 items
Catalog # MILXPD20047S0
Supplier:  Merck
47mm Petri dish with pad, sterile, to be filled with liquid media for microbial culture 1 * 150 items
Catalog # MILXPD20047S0
Supplier:  Merck
Frequently Bought Together