Order Entry
Netherlands
ContactUsLinkComponent
The Micro Bicinchoninic Acid (BCA) Protein Assay is a highly sensitive colorimetric assay that is compatible with detergent solubili zed protein solutions. This modification of the Bicinchoninic Acid (BCA) Protein Assay is suitable for dilute protein 1 * 250 mL
Catalog # GENO786-861
Supplier:  G-Biosciences
The Micro Bicinchoninic Acid (BCA) Protein Assay is a highly sensitive colorimetric assay that is compatible with detergent solubili zed protein solutions. This modification of the Bicinchoninic Acid (BCA) Protein Assay is suitable for dilute protein 1 * 250 mL
Catalog # GENO786-861
Supplier:  G-Biosciences