Order Entry
Netherlands
ContactUsLinkComponent
The Bicinchoninic Acid (BCA) Protein Assay, also known as the Smith Assay, is a highly sensitive colorimetric assay that is compatib le with detergent solubilized protein solutions. 1 * 25 mL
Catalog # GENO786-848
Supplier:  G-Biosciences
The Bicinchoninic Acid (BCA) Protein Assay, also known as the Smith Assay, is a highly sensitive colorimetric assay that is compatib le with detergent solubilized protein solutions. 1 * 25 mL
Catalog # GENO786-848
Supplier:  G-Biosciences