Order Entry
Netherlands
ContactUsLinkComponent
EDC is a heterobifunctional, water-soluble, zero-length carbodiimide crosslinker that is used to couple carboxyl groups to primary a mines. EDC activates carboxyl groups first and forms amine reactive O-acylisourea imtermediate that spontaneously rea 1 * 50 mg
Catalog # GENOBC25-50
Supplier:  G-Biosciences
EDC is a heterobifunctional, water-soluble, zero-length carbodiimide crosslinker that is used to couple carboxyl groups to primary a mines. EDC activates carboxyl groups first and forms amine reactive O-acylisourea imtermediate that spontaneously rea 1 * 50 mg
Catalog # GENOBC25-50
Supplier:  G-Biosciences