Order Entry
Netherlands
ContactUsLinkComponent
Our Immobilized Boronic Acid is designed for the isolation of ribonucleotide and oligonucleotide RNA.The resin consists of boronic a cid covalently linked to a polyacrylamide support that offers simple isolation of small molecular weight compounds th 1 * 2 mL
Catalog # GENO786-823
Supplier:  G-Biosciences
Our Immobilized Boronic Acid is designed for the isolation of ribonucleotide and oligonucleotide RNA.The resin consists of boronic a cid covalently linked to a polyacrylamide support that offers simple isolation of small molecular weight compounds th 1 * 2 mL
Catalog # GENO786-823
Supplier:  G-Biosciences