Buffer Solution pH 10 for volumetric analysis 1 * 1.000 mL
Catalog # APPC281730.1211
Supplier: PanReac AppliChem
Buffer Solution pH 10 for volumetric analysis 1 * 1.000 mL
Catalog # APPC281730.1211
Supplier: PanReac AppliChem
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck