Recombinant protein of human survival of motor neuron 1, telomeric (SMN1), transcript variant d 1 * 1 EA
Catalog # ORIGTP321367
Supplier: OriGene
Recombinant protein of human survival of motor neuron 1, telomeric (SMN1), transcript variant d 1 * 1 EA
Catalog # ORIGTP321367
Supplier: OriGene
Frequently Bought Together
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrilhandschoenen
Supplier: VWR Collection
VWR®, Transferpipetten, Hoge Prestaties
Supplier: VWR Collection
VWR®, Serologische pipetten, standaardreeks
Supplier: VWR Collection
Nitril handschoenen, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrilhandschoenen
Supplier: VWR Collection
VWR®, gezichtstissues
Supplier: VWR Collection
pH-teststrips, MQuant®, Supelco®
Supplier: Merck