Order Entry
Netherlands
ContactUsLinkComponent
Peristaltic pump tubing, PVC Solvaflex, orange/yellow tabs, flared ends, 0.51 mm id, for use with SPS 3 autosampler when pumping low volatile organic samples, 12/pk 1 * 12 items
Catalog # HEWL3710069000
Supplier:  Agilent
Peristaltic pump tubing, PVC Solvaflex, orange/yellow tabs, flared ends, 0.51 mm id, for use with SPS 3 autosampler when pumping low volatile organic samples, 12/pk 1 * 12 items
Catalog # HEWL3710069000
Supplier:  Agilent
Frequently Bought Together